Product Details

SNP ID
rs3820447
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:149903449 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GATGGATGGGTCCCTCCCAGGGGTG[C/T]GAGGAAGAGGGCTACGTCCAGCATG
Phenotype
MIM: 613181 MIM: 185860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOLA1 PubMed Links

Gene Details

Gene
BOLA1
Gene Name
bolA family member 1
There are no transcripts associated with this gene.

Gene
SV2A
Gene Name
synaptic vesicle glycoprotein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278719.1 2251 UTR 3 NP_001265648.1
NM_014849.4 2251 UTR 3 NP_055664.3
XM_017002992.1 2251 Intron XP_016858481.1

View Full Product Details