Product Details
- SNP ID
-
rs3820654
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:172663801 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- GTAAAATGTCACTGGGGGAAGATCT[C/G]GTGACACGCAGACAGATCAGTAGTC
- Phenotype
-
MIM: 134638
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FASLG
PubMed Links
Gene Details
- Gene
- FASLG
- Gene Name
- Fas ligand
View Full Product Details