Product Details

SNP ID
rs3750328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128735253 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTGACAGCAGCAATTAATGCCCC[C/G]CTCCTGGACTACAAACTCCCTGAGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100506100 PubMed Links

Gene Details

Gene
LOC100506100
Gene Name
uncharacterized LOC100506100
There are no transcripts associated with this gene.

Gene
ZER1
Gene Name
zyg-11 related cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006336.3 Intron NP_006327.2
XM_005251645.2 Intron XP_005251702.1
XM_005251646.2 Intron XP_005251703.1
XM_011518122.2 Intron XP_011516424.1
XM_017014185.1 Intron XP_016869674.1
XM_017014186.1 Intron XP_016869675.1
XM_017014187.1 Intron XP_016869676.1
XM_017014188.1 Intron XP_016869677.1
XM_017014189.1 Intron XP_016869678.1
XM_017014190.1 Intron XP_016869679.1

View Full Product Details