Product Details

SNP ID
rs3756289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131646501 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGCTTTCTTTAATGGCATTTCA[A/G]GAAATAATTCATATGAACCCACCTA
Phenotype
MIM: 610594
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNIP1 PubMed Links

Gene Details

Gene
FNIP1
Gene Name
folliculin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008738.2 Intron NP_001008738.2
NM_133372.2 Intron NP_588613.2

View Full Product Details