Product Details

SNP ID
rs3759096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56153026 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAATACTCATGGAGCCATAGGAG[C/G]GCCCACATCCTCTTGGATTAGGGAA
Phenotype
MIM: 616670 MIM: 609931 MIM: 609930 MIM: 601734
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ESYT1 PubMed Links

Gene Details

Gene
ESYT1
Gene Name
extended synaptotagmin 1
There are no transcripts associated with this gene.

Gene
LOC107987175
Gene Name
uncharacterized LOC107987175
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017020294.1 Intron XP_016875783.1
Gene
MYL6
Gene Name
myosin light chain 6
There are no transcripts associated with this gene.

Gene
MYL6B
Gene Name
myosin light chain 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199629.1 Intron NP_001186558.1
NM_002475.4 Intron NP_002466.1
Gene
SMARCC2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
There are no transcripts associated with this gene.

View Full Product Details