Product Details

SNP ID
rs3765484
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:40258425 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CCTAGACAGGGTCCAACCTGACCCC[C/G]ATACCCGGCCAGCCCTGGAGTAGCC
Phenotype
MIM: 606480
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMCO2 PubMed Links

Gene Details

Gene
TMCO2
Gene Name
transmembrane and coiled-coil domains 2
There are no transcripts associated with this gene.

Gene
ZMPSTE24
Gene Name
zinc metallopeptidase STE24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005857.4 Intron NP_005848.2

View Full Product Details