Product Details

SNP ID
rs3768425
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:226990426 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GAGTAACAAAGTCATAAGGAAAGAG[A/G]TTTTAAAATGGGCTCCATGAAATGC
Phenotype
MIM: 603412 MIM: 606980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC42BPA PubMed Links

Gene Details

Gene
CDC42BPA
Gene Name
CDC42 binding protein kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003607.3 10249 UTR 3 NP_003598.2
NM_014826.4 10249 UTR 3 NP_055641.3
XM_005273317.3 10249 Intron XP_005273374.1
XM_005273318.3 10249 Intron XP_005273375.1
XM_005273320.3 10249 Intron XP_005273377.1
XM_005273321.3 10249 UTR 3 XP_005273378.1
XM_005273322.3 10249 UTR 3 XP_005273379.1
XM_005273324.3 10249 UTR 3 XP_005273381.1
XM_006711834.3 10249 Intron XP_006711897.1
XM_006711835.3 10249 Intron XP_006711898.1
XM_011544306.2 10249 Intron XP_011542608.1
XM_011544307.2 10249 Intron XP_011542609.1
XM_011544308.2 10249 UTR 3 XP_011542610.1
XM_011544309.2 10249 Intron XP_011542611.1
XM_017002578.1 10249 UTR 3 XP_016858067.1
XM_017002579.1 10249 Intron XP_016858068.1
XM_017002580.1 10249 Intron XP_016858069.1
XM_017002581.1 10249 Intron XP_016858070.1
XM_017002582.1 10249 Intron XP_016858071.1
Gene
COQ8A
Gene Name
coenzyme Q8A
There are no transcripts associated with this gene.

View Full Product Details