Product Details
- SNP ID
-
rs3769826
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:201238326 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGGAATATATGGGATATCATTTA[A/C]ATTTTCTAAAGGTTATCTACAAGAT
- Phenotype
-
MIM: 601762
MIM: 601763
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CASP10
PubMed Links
Gene Details
- Gene
- CASP10
- Gene Name
- caspase 10
There are no transcripts associated with this gene.
- Gene
- CASP8
- Gene Name
- caspase 8
View Full Product Details