Product Details

SNP ID
rs3203916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92714676 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCATGAAACTATGCCAAGTGCA[A/G]AGCCATCCTCTGAGGTCGTACCTAT
Phenotype
MIM: 609797
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BICD2 PubMed Links

Gene Details

Gene
BICD2
Gene Name
BICD cargo adaptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003800.1 3103 UTR 3 NP_001003800.1
NM_015250.3 3103 Intron NP_056065.1
XM_017014551.1 3103 Intron XP_016870040.1

View Full Product Details