Product Details

SNP ID
rs3787016
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1090804 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACCCTGTCCTCCATGCACTAAT[A/G]GGAACACCTGCCCTGGGCCCCAGAT
Phenotype
MIM: 601155 MIM: 180664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP45 PubMed Links

Gene Details

Gene
ARHGAP45
Gene Name
Rho GTPase activating protein 45
There are no transcripts associated with this gene.

Gene
POLR2E
Gene Name
RNA polymerase II subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316323.1 Intron NP_001303252.1
NM_001316324.1 Intron NP_001303253.1
NM_002695.3 Intron NP_002686.2
XM_011528070.2 Intron XP_011526372.1
XM_017026883.1 Intron XP_016882372.1

View Full Product Details