Product Details

SNP ID
rs3794486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:105700054 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCACATCAAACCACCATTTTGT[A/G]TAAGACTTCCTGACTACAGTAATTT
Phenotype
MIM: 611236
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM38B PubMed Links

Gene Details

Gene
TMEM38B
Gene Name
transmembrane protein 38B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018112.2 Intron NP_060582.1
XM_005252075.2 Intron XP_005252132.1
XM_005252076.2 Intron XP_005252133.1
XM_005252077.3 Intron XP_005252134.1
XM_011518829.1 Intron XP_011517131.1
XM_011518831.2 Intron XP_011517133.1
XM_011518832.2 Intron XP_011517134.1
XM_011518833.2 Intron XP_011517135.1

View Full Product Details