Product Details
- SNP ID
-
rs3800583
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:128833148 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCCTCACCACCTTGTCCTGGCCC[C/T]CCGTTGCCCCTCGTTCCCTCCGAAC
- Phenotype
-
MIM: 102565
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLNC
PubMed Links
Gene Details
- Gene
- FLNC
- Gene Name
- filamin C
View Full Product Details