Product Details

SNP ID
rs3803415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65896417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCATGGCAGGCTTCCAGTTTCT[A/G]TCAAGGGTTTCAGAACTCTCATACT
Phenotype
MIM: 612454 MIM: 605570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEGF11 PubMed Links

Gene Details

Gene
MEGF11
Gene Name
multiple EGF like domains 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032445.2 5222 UTR 3 NP_115821.2
XM_017022670.1 5222 UTR 3 XP_016878159.1
XM_017022671.1 5222 UTR 3 XP_016878160.1
XM_017022672.1 5222 UTR 3 XP_016878161.1
XM_017022673.1 5222 Intron XP_016878162.1
XM_017022674.1 5222 UTR 3 XP_016878163.1
XM_017022675.1 5222 UTR 3 XP_016878164.1
Gene
RAB11A
Gene Name
RAB11A, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details