Product Details
- SNP ID
-
rs3806439
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
28
- Location
-
Chr.1:228143802 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- AAGCGCAGGGGCAAATTCTGCGGGT[A/G]GGAGAGCCAGGAGGGCATGGGGTGC
- Phenotype
-
MIM: 608803
MIM: 139270
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GJC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112179347] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GJC2
- Gene Name
- gap junction protein gamma 2
There are no transcripts associated with this gene.
- Gene
- GUK1
- Gene Name
- guanylate kinase 1
View Full Product Details