Product Details

SNP ID
rs3808172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:118232955 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGTGGGCTTAAAGCTATTTAGC[C/T]CTGTGTTATGTTTGGATTGAGGGCT
Phenotype
MIM: 610731
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD7 PubMed Links

Gene Details

Gene
ANKRD7
Gene Name
ankyrin repeat domain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019644.3 Intron NP_062618.2

View Full Product Details