Product Details

SNP ID
rs3809563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:70855937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAAAGAGCTGCAGCAGCCACC[A/G]TGCCTTCCAGTTTGATAATTTCGGT
Phenotype
MIM: 611300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LARP6 PubMed Links

Gene Details

Gene
LARP6
Gene Name
La ribonucleoprotein domain family member 6
There are no transcripts associated with this gene.

Gene
LRRC49
Gene Name
leucine rich repeat containing 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199017.2 Intron NP_001185946.1
NM_001199018.2 Intron NP_001185947.1
NM_001284357.1 Intron NP_001271286.1
NM_017691.4 Intron NP_060161.2
XM_005254492.4 Intron XP_005254549.1
XM_011521715.2 Intron XP_011520017.1
XM_011521717.2 Intron XP_011520019.1
XM_011521718.2 Intron XP_011520020.1
XM_017022357.1 Intron XP_016877846.1
XM_017022358.1 Intron XP_016877847.1
XM_017022359.1 Intron XP_016877848.1

View Full Product Details