Product Details

SNP ID
rs3810293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47228539 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGGGTTAGGACCCAGATGGAGAA[C/G]AGTGGAGGTGTGTGTGCATGCGGAG
Phenotype
MIM: 605854
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BBC3 PubMed Links

Gene Details

Gene
BBC3
Gene Name
BCL2 binding component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127240.2 Intron NP_001120712.1
NM_001127241.2 Intron NP_001120713.1
NM_001127242.2 Intron NP_001120714.1
NM_014417.4 Intron NP_055232.1
XM_006723141.3 Intron XP_006723204.1
XM_011526722.2 Intron XP_011525024.1
Gene
MIR3190
Gene Name
microRNA 3190
There are no transcripts associated with this gene.

Gene
MIR3191
Gene Name
microRNA 3191
There are no transcripts associated with this gene.

View Full Product Details