Product Details

SNP ID
rs3811404
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:151170913 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GAATGCTAACATCAGGGGAAGGGAA[C/T]GCTGACCTGATTGTCTACACGGAGC
Phenotype
MIM: 608095 MIM: 605834 MIM: 600607
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYSMD1 PubMed Links

Gene Details

Gene
LYSMD1
Gene Name
LysM domain containing 1
There are no transcripts associated with this gene.

Gene
SCNM1
Gene Name
sodium channel modifier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204856.1 Intron NP_001191785.1
NM_024041.3 Intron NP_076946.1
Gene
TMOD4
Gene Name
tropomodulin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013353.2 Intron NP_037485.2
XM_011509449.1 Intron XP_011507751.1
XM_017001089.1 Intron XP_016856578.1
XM_017001090.1 Intron XP_016856579.1
Gene
TNFAIP8L2-SCNM1
Gene Name
TNFAIP8L2-SCNM1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204848.1 Intron NP_001191777.1
Gene
VPS72
Gene Name
vacuolar protein sorting 72 homolog
There are no transcripts associated with this gene.

View Full Product Details