Product Details
- SNP ID
-
rs3813903
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31808307 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACACAAAACATCAAGGGATGATAA[C/T]AAGACAATATGTAATGAATGTGGAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VN1R3
PubMed Links
Gene Details
- Gene
- VN1R3
- Gene Name
- vomeronasal 1 receptor 3 (gene/pseudogene)
There are no transcripts associated with this gene.
View Full Product Details