Product Details

SNP ID
rs3814351
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:152720386 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CAGTGATGTAGAACCTCATCACTTT[C/G]CCTCTGTGCTTTGCTTCTCTACCAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf68 PubMed Links

Gene Details

Gene
C1orf68
Gene Name
chromosome 1 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024679.2 865 UTR 3 NP_001019850.1

View Full Product Details