Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042550.1 | 44 | Intron | NP_001036015.1 | ||
NM_001042551.1 | 44 | UTR 5 | NP_001036016.1 | ||
NM_001265602.1 | 44 | UTR 5 | NP_001252531.1 | ||
NM_006444.2 | 44 | UTR 5 | NP_006435.2 | ||
XM_006716933.3 | 44 | UTR 5 | XP_006716996.1 | ||
XM_011518148.2 | 44 | Intron | XP_011516450.1 | ||
XM_011518149.2 | 44 | Intron | XP_011516451.1 | ||
XM_011518150.2 | 44 | UTR 5 | XP_011516452.1 | ||
XM_011518151.2 | 44 | UTR 5 | XP_011516453.1 | ||
XM_011518152.2 | 44 | UTR 5 | XP_011516454.1 | ||
XM_011518153.1 | 44 | UTR 5 | XP_011516455.1 | ||
XM_017014206.1 | 44 | Intron | XP_016869695.1 | ||
XM_017014207.1 | 44 | UTR 5 | XP_016869696.1 | ||
XM_017014208.1 | 44 | Intron | XP_016869697.1 | ||
XM_017014209.1 | 44 | Intron | XP_016869698.1 | ||
XM_017014210.1 | 44 | UTR 5 | XP_016869699.1 | ||
XM_017014211.1 | 44 | UTR 5 | XP_016869700.1 | ||
XM_017014212.1 | 44 | Intron | XP_016869701.1 |