Product Details

SNP ID
rs3820310
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:180158562 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GAAGGCATTTTGTCCATCCAGTGAC[A/G]TCACTGCCCAGAAACTGTCAGCAAG
Phenotype
MIM: 603120
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
QSOX1 PubMed Links

Gene Details

Gene
QSOX1
Gene Name
quiescin sulfhydryl oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004128.2 Intron NP_001004128.1
NM_002826.4 Intron NP_002817.2

View Full Product Details