Product Details

SNP ID
rs3825546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95533483 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGTCATTATCACCGTGGCAACAG[C/T]GACCAGAAACTATCAGAGGAAAATT
Phenotype
MIM: 609588
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLRX5 PubMed Links

Gene Details

Gene
GLRX5
Gene Name
glutaredoxin 5
There are no transcripts associated with this gene.

Gene
SCARNA13
Gene Name
small Cajal body-specific RNA 13
There are no transcripts associated with this gene.

Gene
SNHG10
Gene Name
small nucleolar RNA host gene 10
There are no transcripts associated with this gene.

View Full Product Details