Product Details

SNP ID
rs3829686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1214658 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGGCCCCCACGAGGCTCTGAGGCT[A/G]CATGCAGGTTGAGCTCCTGGCCTCA
Phenotype
MIM: 602216
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STK11 PubMed Links

Gene Details

Gene
STK11
Gene Name
serine/threonine kinase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000455.4 Intron NP_000446.1
XM_005259617.2 Intron XP_005259674.1
XM_005259618.3 Intron XP_005259675.1
XM_011528209.1 Intron XP_011526511.1

View Full Product Details