Product Details

SNP ID
rs3206108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:53516420 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCATCCCAGCATAAGACCTGAGG[C/G]TGGTCCCATGCTCAAAAATGTTTCT
Phenotype
MIM: 613214
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WDR72 PubMed Links

Gene Details

Gene
WDR72
Gene Name
WD repeat domain 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277176.1 4827 UTR 3 NP_001264105.1
NM_182758.3 4827 UTR 3 NP_877435.3
XM_011521433.2 4827 UTR 3 XP_011519735.1
XM_011521435.2 4827 UTR 3 XP_011519737.1
XM_011521436.2 4827 Intron XP_011519738.1
XM_011521437.2 4827 Intron XP_011519739.1
XM_017022061.1 4827 UTR 3 XP_016877550.1

View Full Product Details