Product Details

SNP ID
rs3210392
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:63549108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGAGCAAAGGGCAGTAGGCGG[A/C]GCTTCGCCAAGGCTGGCGCAGTCGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DLEU2L PubMed Links

Gene Details

Gene
DLEU2L
Gene Name
deleted in lymphocytic leukemia 2-like
There are no transcripts associated with this gene.

Gene
EFCAB7
Gene Name
EF-hand calcium binding domain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032437.3 Intron NP_115813.2
XM_005271274.3 Intron XP_005271331.1
XM_006710976.3 Intron XP_006711039.1
XM_006710977.1 Intron XP_006711040.1
XM_011542301.2 Intron XP_011540603.1
XM_017002548.1 Intron XP_016858037.1

View Full Product Details