Product Details

SNP ID
rs3210572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88523586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCTCCTCAGTCTTCTTGCCCC[C/G]GTCGGCCAAAGGCCGCGACCCCGAC
Phenotype
MIM: 610662
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929134 PubMed Links

Gene Details

Gene
LOC101929134
Gene Name
uncharacterized LOC101929134
There are no transcripts associated with this gene.

Gene
PIGY
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Y
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042616.2 216 UTR 5 NP_001036081.1
Gene
PYURF
Gene Name
PIGY upstream reading frame
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032906.4 216 Silent Mutation CGG,GGG R39G NP_116295.1

View Full Product Details