Product Details

SNP ID
rs3212292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:12739604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGGGATTTTTCTCCTGGACAAC[A/T]TGATCATCTTTTTGATTGCTTTAAG
Phenotype
MIM: 400005
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
USP9Y PubMed Links

Gene Details

Gene
USP9Y
Gene Name
ubiquitin specific peptidase 9, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004654.3 2342 Missense Mutation CAT,CTT H466L NP_004645.2
XM_017030078.1 2342 Missense Mutation CAT,CTT H466L XP_016885567.1

View Full Product Details