Product Details

SNP ID
rs3732750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:55468223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAAGTACTGTCATTTCTAATAG[A/G]CATGGGTTTCCATTCTGCAGAATGA
Phenotype
MIM: 164975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT5A PubMed Links

Gene Details

Gene
WNT5A
Gene Name
Wnt family member 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256105.1 3221 UTR 3 NP_001243034.1
NM_003392.4 3221 UTR 3 NP_003383.2
XM_006713324.1 3221 UTR 3 XP_006713387.1
XM_011534085.2 3221 UTR 3 XP_011532387.1
XM_011534086.2 3221 UTR 3 XP_011532388.1
XM_011534087.2 3221 UTR 3 XP_011532389.1
XM_011534088.2 3221 UTR 3 XP_011532390.1
XM_011534089.1 3221 UTR 3 XP_011532391.1
XM_017007127.1 3221 UTR 3 XP_016862616.1
XM_017007128.1 3221 UTR 3 XP_016862617.1

View Full Product Details