Product Details

SNP ID
rs3733763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10680360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGAGCCTGTCTGGGCTGAGGC[C/G]ATGCTGGGCTTGCCGTGCCGAGATC
Phenotype
MIM: 600954
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DAP PubMed Links

Gene Details

Gene
DAP
Gene Name
death associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291963.1 1169 UTR 3 NP_001278892.1
NM_004394.2 1169 UTR 3 NP_004385.1

View Full Product Details