Product Details

SNP ID
rs3735137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39951848 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTCGCAGTCCCTACAGCCCTGTG[C/T]TCAGGTGAGTTCTGCCGTTCTGCCT
Phenotype
MIM: 603309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK13 PubMed Links

Gene Details

Gene
CDK13
Gene Name
cyclin dependent kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003718.4 2348 Missense Mutation CTC,TTC L403F NP_003709.3
NM_031267.3 2348 Missense Mutation CTC,TTC L403F NP_112557.2
XM_011515597.2 2348 Missense Mutation CTC,TTC L403F XP_011513899.1
XM_017012750.1 2348 Missense Mutation CTC,TTC L403F XP_016868239.1
XM_017012751.1 2348 Missense Mutation CTC,TTC L403F XP_016868240.1

View Full Product Details