Product Details

SNP ID
rs3737638
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:2324085 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GCACAGCCGGCGATGGACTTGGGCC[T/C]TGTCCACCTACCTAGGTGTGCTGCC
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MORN1 PubMed Links

Gene Details

Gene
MORN1
Gene Name
MORN repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301060.1 Intron NP_001287989.1
NM_024848.2 Intron NP_079124.1
XM_005244798.2 Intron XP_005244855.1
XM_011542169.1 Intron XP_011540471.1
XM_011542170.2 Intron XP_011540472.1
XM_011542172.2 Intron XP_011540474.1
XM_011542173.2 Intron XP_011540475.1
XM_011542174.2 Intron XP_011540476.1
XM_011542176.1 Intron XP_011540478.1
XM_011542177.1 Intron XP_011540479.1
XM_017002365.1 Intron XP_016857854.1

View Full Product Details