Product Details
- SNP ID
-
rs3737967
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
36
- Location
-
Chr.1:11787392 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- CCTCTCTGGCTATTCCTTCAGACCC[A/G]CAGCTGCTGGACACAGGCCACAGAG
- Phenotype
-
MIM: 607093
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf167
PubMed Links
Gene Details
- Gene
- C1orf167
- Gene Name
- chromosome 1 open reading frame 167
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001010881.1 |
3869 |
Missense Mutation |
CAC,CGC |
H1191R |
NP_001010881.1 |
XM_011541269.2 |
3869 |
Missense Mutation |
CAC,CGC |
H1236R |
XP_011539571.2 |
XM_011541271.2 |
3869 |
Missense Mutation |
CAC,CGC |
H1218R |
XP_011539573.2 |
XM_011541272.2 |
3869 |
Missense Mutation |
CAC,CGC |
H1236R |
XP_011539574.1 |
XM_011541274.2 |
3869 |
Missense Mutation |
CAC,CGC |
H1191R |
XP_011539576.2 |
XM_011541275.2 |
3869 |
Missense Mutation |
CAC,CGC |
H1191R |
XP_011539577.2 |
XM_011541276.2 |
3869 |
Missense Mutation |
CAC,CGC |
H1236R |
XP_011539578.1 |
XM_011541277.2 |
3869 |
Missense Mutation |
CAC,CGC |
H1236R |
XP_011539579.1 |
XM_011541278.2 |
3869 |
Missense Mutation |
CAC,CGC |
H1236R |
XP_011539580.1 |
XM_011541280.2 |
3869 |
Missense Mutation |
CAC,CGC |
H663R |
XP_011539582.2 |
XM_011541281.2 |
3869 |
Missense Mutation |
CAC,CGC |
H663R |
XP_011539583.2 |
XM_017001062.1 |
3869 |
Missense Mutation |
CAC,CGC |
H1236R |
XP_016856551.1 |
XM_017001063.1 |
3869 |
Missense Mutation |
CAC,CGC |
H1236R |
XP_016856552.1 |
XM_017001064.1 |
3869 |
Missense Mutation |
CAC,CGC |
H1236R |
XP_016856553.1 |
- Gene
- LOC102724659
- Gene Name
- uncharacterized LOC102724659
There are no transcripts associated with this gene.
- Gene
- MTHFR
- Gene Name
- methylenetetrahydrofolate reductase (NAD(P)H)
View Full Product Details