Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001127714.2 | 11006 | UTR 3 | NP_001121186.1 | ||
NM_024503.4 | 11006 | UTR 3 | NP_078779.2 | ||
XM_011541884.2 | 11006 | UTR 3 | XP_011540186.1 | ||
XM_011541885.2 | 11006 | Intron | XP_011540187.1 | ||
XM_011541886.2 | 11006 | Intron | XP_011540188.1 | ||
XM_011541887.2 | 11006 | Intron | XP_011540189.1 | ||
XM_017001992.1 | 11006 | UTR 3 | XP_016857481.1 | ||
XM_017001993.1 | 11006 | UTR 3 | XP_016857482.1 | ||
XM_017001994.1 | 11006 | UTR 3 | XP_016857483.1 |