Product Details

SNP ID
rs3738819
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:183251443 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GAAAGAGACATTTCCCAGCCCAACA[C/G]ACAATACATAATTCCAATTAGCAAA
Phenotype
MIM: 150292 MIM: 608701
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LAMC2 PubMed Links

Gene Details

Gene
LAMC2
Gene Name
laminin subunit gamma 2
There are no transcripts associated with this gene.

Gene
NMNAT2
Gene Name
nicotinamide nucleotide adenylyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015039.3 2354 UTR 3 NP_055854.1
NM_170706.3 2354 UTR 3 NP_733820.1

View Full Product Details