Product Details

SNP ID
rs3740518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97332615 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTACATCAATAGCTACATCTGGAG[A/G]ATGAACAAGCCATTGAATCTTTTAA
Phenotype
MIM: 605006
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRAT2 PubMed Links

Gene Details

Gene
FRAT2
Gene Name
frequently rearranged in advanced T-cell lymphomas 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012083.2 2087 UTR 3 NP_036215.1

View Full Product Details