Product Details

SNP ID
rs3740538
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124621221 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCGGCCACACCGGGCATGTGGTG[A/C]CAGGACCCCCCACGCCCAGTGGTAC
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
FAM53B PubMed Links

Gene Details

Gene
FAM53B
Gene Name
family with sequence similarity 53 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014661.3 3813 UTR 3 NP_055476.3
Gene
LHPP
Gene Name
phospholysine phosphohistidine inorganic pyrophosphate phosphatase
There are no transcripts associated with this gene.

View Full Product Details