Product Details

SNP ID
rs3741918
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6535090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCCGGAGCCTCCTTCCCCTAG[A/T]CCCCAGAAACAGGAGGTCCCTACTC
Phenotype
MIM: 138400 MIM: 610495 MIM: 615638
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
GAPDH PubMed Links
Additional Information
For this assay, SNP(s) [rs45524841] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GAPDH
Gene Name
glyceraldehyde-3-phosphate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256799.2 Intron NP_001243728.1
NM_001289745.1 Intron NP_001276674.1
NM_001289746.1 Intron NP_001276675.1
NM_002046.5 Intron NP_002037.2
Gene
IFFO1
Gene Name
intermediate filament family orphan 1
There are no transcripts associated with this gene.

Gene
NCAPD2
Gene Name
non-SMC condensin I complex subunit D2
There are no transcripts associated with this gene.

View Full Product Details