Product Details

SNP ID
rs3742297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52703932 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGAAGGTGTAATCTGTAAAGGG[C/T]TTGATCTTGTTTACCCTGTCATTCA
Phenotype
MIM: 605147 MIM: 604098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LECT1 PubMed Links

Gene Details

Gene
LECT1
Gene Name
leukocyte cell derived chemotaxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011705.1 Intron NP_001011705.1
NM_007015.2 Intron NP_008946.1
XM_011534897.2 Intron XP_011533199.1
XM_011534898.2 Intron XP_011533200.1
XM_011534899.2 Intron XP_011533201.1
XM_011534900.2 Intron XP_011533202.1
Gene
SUGT1
Gene Name
SGT1 homolog, MIS12 kinetochore complex assembly cochaperone
There are no transcripts associated with this gene.

View Full Product Details