Product Details

SNP ID
rs3746317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17556110 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCGCGTGCTTCCTGCTCGCTACCC[C/T]CATCGGGACGGTTCTACGCATGCCC
Phenotype
MIM: 609967
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COLGALT1 PubMed Links

Gene Details

Gene
COLGALT1
Gene Name
collagen beta(1-O)galactosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024656.3 2805 Intron NP_078932.2
XM_005260080.4 2805 Intron XP_005260137.1
XM_011528297.2 2805 Intron XP_011526599.1
XM_011528298.2 2805 Intron XP_011526600.1
XM_011528299.2 2805 Intron XP_011526601.1
Gene
FAM129C
Gene Name
family with sequence similarity 129 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098524.1 2805 Intron NP_001091994.1
NM_001321826.1 2805 Intron NP_001308755.1
NM_001321827.1 2805 Intron NP_001308756.1
NM_001321828.1 2805 Intron NP_001308757.1
NM_173544.4 2805 Intron NP_775815.2
XM_005259813.3 2805 Intron XP_005259870.1
XM_005259814.3 2805 Intron XP_005259871.1
XM_011527781.2 2805 Intron XP_011526083.1
XM_011527786.2 2805 UTR 3 XP_011526088.1
XM_011527787.2 2805 Intron XP_011526089.1
XM_011527789.1 2805 Intron XP_011526091.1
XM_017026453.1 2805 Intron XP_016881942.1
XM_017026454.1 2805 Intron XP_016881943.1
XM_017026455.1 2805 Intron XP_016881944.1
XM_017026456.1 2805 Intron XP_016881945.1
XM_017026457.1 2805 Intron XP_016881946.1

View Full Product Details