Product Details

SNP ID
rs3748015
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.1:94532704 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
TCCAATTTAGGCATATTGAACCCTA[C/T]TGAGCCCCTTTAGAAGATATCTGCA
Phenotype
MIM: 134390
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
F3 PubMed Links

Gene Details

Gene
F3
Gene Name
coagulation factor III, tissue factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178096.1 Intron NP_001171567.1
NM_001993.4 Intron NP_001984.1

View Full Product Details