Product Details

SNP ID
rs3749035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:170816270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTGGCCAGGTGTGGTACTTTG[A/C]TGGGGCCCAGGGCTGGATGAGGAAA
Phenotype
MIM: 605363
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GAD1 PubMed Links

Gene Details

Gene
GAD1
Gene Name
glutamate decarboxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000817.2 Intron NP_000808.2
NM_013445.3 Intron NP_038473.2
XM_005246444.2 Intron XP_005246501.1
XM_011510922.1 Intron XP_011509224.1
XM_017003756.1 Intron XP_016859245.1
XM_017003757.1 Intron XP_016859246.1
XM_017003758.1 Intron XP_016859247.1

View Full Product Details