Product Details

SNP ID
rs3749210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122426250 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCAAAAGCGATACAAATTAATAT[A/G]TGCCCCTCTCCTTTCTGCTCTAAGG
Phenotype
MIM: 600686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KPNA1 PubMed Links

Gene Details

Gene
KPNA1
Gene Name
karyopherin subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002264.3 4137 UTR 3 NP_002255.3
XM_005247437.3 4137 UTR 3 XP_005247494.1
XM_005247439.3 4137 UTR 3 XP_005247496.1
XM_011512795.2 4137 UTR 3 XP_011511097.1
XM_011512797.2 4137 UTR 3 XP_011511099.1
XM_017006363.1 4137 UTR 3 XP_016861852.1
XM_017006364.1 4137 UTR 3 XP_016861853.1
XM_017006365.1 4137 UTR 3 XP_016861854.1
Gene
LOC102723582
Gene Name
uncharacterized LOC102723582
There are no transcripts associated with this gene.

View Full Product Details