Product Details

SNP ID
rs3750743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49155463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCTAAGAGGGTGAAGCTAATC[A/G]TTCTACAAAGTAGTGCCTAACTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf128 PubMed Links

Gene Details

Gene
C10orf128
Gene Name
chromosome 10 open reading frame 128
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010863.2 1004 UTR 3 NP_001010863.1
NM_001288740.1 1004 UTR 3 NP_001275669.1
NM_001288741.1 1004 UTR 3 NP_001275670.1
NM_001288742.1 1004 UTR 3 NP_001275671.1
NM_001288743.1 1004 UTR 3 NP_001275672.1
XM_011539378.2 1004 Intron XP_011537680.1
XM_011539379.1 1004 UTR 3 XP_011537681.1
XM_011539380.1 1004 UTR 3 XP_011537682.1
XM_011539382.1 1004 Intron XP_011537684.1
XM_011539383.2 1004 UTR 3 XP_011537685.1
XM_011539384.1 1004 UTR 3 XP_011537686.1
XM_011539385.1 1004 UTR 3 XP_011537687.1
XM_017015792.1 1004 Intron XP_016871281.1
XM_017015793.1 1004 Intron XP_016871282.1
XM_017015794.1 1004 Intron XP_016871283.1
XM_017015795.1 1004 Intron XP_016871284.1
XM_017015796.1 1004 Intron XP_016871285.1
XM_017015797.1 1004 Intron XP_016871286.1
Gene
FAM170B-AS1
Gene Name
FAM170B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details