Product Details

SNP ID
rs3754436
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:110212757 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CAAAGCTGAGACCACAATCCGGCCT[C/T]CTTTCCAGGTTTAAGAAAGGCATTC
Phenotype
MIM: 176265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNC4 PubMed Links

Gene Details

Gene
KCNC4
Gene Name
potassium voltage-gated channel subfamily C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039574.2 Intron NP_001034663.1
NM_004978.4 Intron NP_004969.2
XM_006710625.3 Intron XP_006710688.1
XM_006710627.3 Intron XP_006710690.1
XM_011541401.2 Intron XP_011539703.1
XM_011541402.1 Intron XP_011539704.1
XM_011541403.1 Intron XP_011539705.1
XM_011541404.1 Intron XP_011539706.1
Gene
KCNC4-AS1
Gene Name
KCNC4 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details