Product Details

SNP ID
rs3754624
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:224769095 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTCACTCCCAGATTTATGATTT[C/T]CCCCATTCTATCAAGTCATCTCTCC
Phenotype
MIM: 611518
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DOCK10 PubMed Links

Gene Details

Gene
DOCK10
Gene Name
dedicator of cytokinesis 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290263.1 7875 Intron NP_001277192.1
NM_014689.2 7875 Intron NP_055504.2
XM_005246673.4 7875 Intron XP_005246730.1
XM_006712617.3 7875 Intron XP_006712680.1
XM_017004441.1 7875 UTR 3 XP_016859930.1
XM_017004442.1 7875 UTR 3 XP_016859931.1
XM_017004443.1 7875 UTR 3 XP_016859932.1
XM_017004444.1 7875 UTR 3 XP_016859933.1
XM_017004445.1 7875 UTR 3 XP_016859934.1
XM_017004446.1 7875 UTR 3 XP_016859935.1
XM_017004447.1 7875 Intron XP_016859936.1

View Full Product Details