Product Details
- SNP ID
-
rs3760144
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:38801179 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGTAAGGTTACTACACGGTAAGAA[A/G]TGAAGCCGCAAAATAAAAACTTCCA
- Phenotype
-
MIM: 603261
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CWC25
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116061874] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CWC25
- Gene Name
- CWC25 spliceosome associated protein homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017748.4 |
2358 |
UTR 3 |
|
|
NP_060218.1 |
- Gene
- PIP4K2B
- Gene Name
- phosphatidylinositol-5-phosphate 4-kinase type 2 beta
There are no transcripts associated with this gene.
View Full Product Details