Product Details

SNP ID
rs3762053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94490326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTGGTCAGCTGACTGACAATTT[G/T]CTCTTCTGCAAATTGGGAGTACTAA
Phenotype
MIM: 616447
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1429 PubMed Links

Gene Details

Gene
KIAA1429
Gene Name
KIAA1429
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015496.4 Intron NP_056311.2
NM_183009.2 Intron NP_892121.1
XM_017013306.1 Intron XP_016868795.1
XM_017013307.1 Intron XP_016868796.1
XM_017013308.1 Intron XP_016868797.1

View Full Product Details