Product Details

SNP ID
rs3764878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67392137 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGCCGCGGAGCCGCCAGCCTAAC[C/T]CCCTTCCGCTCTTCTCCCCGCAGTG
Phenotype
MIM: 176875 MIM: 603761
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100130987 PubMed Links

Gene Details

Gene
LOC100130987
Gene Name
uncharacterized LOC100130987
There are no transcripts associated with this gene.

Gene
PPP1CA
Gene Name
protein phosphatase 1 catalytic subunit alpha
There are no transcripts associated with this gene.

Gene
RAD9A
Gene Name
RAD9 checkpoint clamp component A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243224.1 Intron NP_001230153.1
NM_004584.2 Intron NP_004575.1
XM_006718652.3 Intron XP_006718715.1
XM_017018097.1 Intron XP_016873586.1
XM_017018098.1 Intron XP_016873587.1

View Full Product Details