Product Details

SNP ID
rs3770521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216115437 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGATTACCTTGCCTTAAGATGGAT[A/G]CTTGTCTAGGCGGCAGATACCTGCC
Phenotype
MIM: 194364
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XRCC5 PubMed Links

Gene Details

Gene
XRCC5
Gene Name
X-ray repair cross complementing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021141.3 Intron NP_066964.1

View Full Product Details